Gene Transcripts

Gene Symbol
Art2b
Entrez Gene ID
11872
Gene Name
ADP-ribosyltransferase 2b
Gene Aliases
ART2.2, ARTC2, Art, Ly92b, Rt-6, Rt6, Rt6-2
Chromosome Location
Chr. 7: 101575729 - 101585583 on Build GRCm38
UniGene ID
Mm.57008
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019915.2 NP_064299.2 2 692
GenBank mRNA AF016463.1 - 2 692
AF016465.1 - 2 692

View Full Product Details