Gene Transcripts
- Gene Symbol
- Arsb
- Entrez Gene ID
- 11881
- Gene Name
- arylsulfatase B
- Gene Aliases
- 1110007C02Rik, AI480648, As-1, As-1r, As-1s, As-1t, As1, As1-r, As1-s, As1-t, Asr-1, Ast-1
- Chromosome Location
- Chr. 13: 93771023 - 93943016 on Build GRCm38
- UniGene ID
- Mm.300178
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009712.3 |
NP_033842.3
|
5, 6 |
1234 |
|
GenBank mRNA |
AK140163.1 |
- |
5, 6 |
1285 |
|
AK154098.1 |
- |
5, 6 |
1154 |
|
AK158312.1 |
- |
5, 6 |
1235 |
|
AK189106.1 |
- |
1, 2 |
100 |
|
AK190270.1 |
- |
2, 3 |
296 |
|
AK191477.1 |
- |
1, 2 |
100 |
|
AK195452.1 |
- |
1, 2 |
100 |
|
AK198200.1 |
- |
1, 2 |
100 |
|
AK211718.1 |
- |
1, 2 |
246 |
|
AK212274.1 |
- |
1, 2 |
100 |
|
AK212990.1 |
- |
1, 2 |
100 |
|
X92096.1 |
- |
4, 5 |
727 |
|
View Full Product Details