Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007568.5 | NP_031594.1 | 5 | 788 | |
GenBank mRNA | AK046658.1 | - | 4 | 2069 | |
AK050206.1 | - | 5 | 604 | ||
AK076272.1 | - | 5 | 708 | ||
BC119152.1 | - | 5 | 654 | 8733901 | |
BC119154.1 | - | 5 | 654 | 8733903 | |
BC144906.1 | - | 4 | 536 | ||
L08394.1 | - | 5 | 627 |