Gene Transcripts

Gene Symbol
Camp
Entrez Gene ID
12796
Gene Name
cathelicidin antimicrobial peptide
Gene Aliases
CAP18, Cnlp, Cramp, FALL39, MCLP
Chromosome Location
Chr. 9: 109847377 - 109849456 on Build GRCm38
UniGene ID
Mm.3834
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009921.2 NP_034051.2 2 244
GenBank mRNA AK158005.1 - 2 236
BC120752.1 - 2 241 40129675
BC120754.1 - 2 244 40129677
BY582734.1 - 1 45
U43409.1 - 2 237
X94353.1 - 2 236

View Full Product Details