Gene Transcripts

Gene Symbol
Smc1b
Entrez Gene ID
140557
Gene Name
structural maintenance of chromosomes 1B
Gene Aliases
SMC-1B, SMC1beta, Smc1l2
Chromosome Location
Chr. 15: 85064689 - 85131957 on Build GRCm38
UniGene ID
Mm.182737
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080470.1 NP_536718.1 24, 25 3628
GenBank mRNA AF303827.1 - 24, 25 3628
BC129800.1 - 24, 25 3619 40048794

View Full Product Details