Gene Transcripts

Gene Symbol
Vmn1r28
Entrez Gene ID
171198
Gene Name
vomeronasal 1 receptor 28
Gene Aliases
V1rc25
Chromosome Location
Chr. 6: 58262294 - 58266082 on Build GRCm38
UniGene ID
Mm.222640
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134180.1 NP_598941.1 1 619
GenBank mRNA AF394949.1 - 775

View Full Product Details