Gene Transcripts

Gene Symbol
Olfr45
Entrez Gene ID
18344
Gene Name
olfactory receptor 45
Gene Aliases
IB6, MOR253-2
Chromosome Location
Chr. 7: 140690907 - 140691842 on Build GRCm38
UniGene ID
Mm.222838
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146963.1 NP_667174.1 1 578
GenBank mRNA AF102519.1 - 1 386
BC125298.1 - 1 601 40129813
BC125300.1 - 1 601 40129815

View Full Product Details