Gene Transcripts

Gene Symbol
Urod
Entrez Gene ID
22275
Gene Name
uroporphyrinogen decarboxylase
Gene Aliases
AI323803, Uro-d
Chromosome Location
Chr. 4: 116990212 - 116994436 on Build GRCm38
UniGene ID
Mm.46484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009478.3 NP_033504.2 10 1009
GenBank mRNA AK002844.1 - 10 998
AK007470.1 - 10 1000
AK172426.1 - 10 1020
BC008109.1 - 10 1009 3481699

View Full Product Details