Gene Transcripts

Gene Symbol
Rtp1
Entrez Gene ID
239766
Gene Name
receptor transporter protein 1
Gene Aliases
Gm1766, Gm604
Chromosome Location
Chr. 16: 23428601 - 23433960 on Build GRCm38
UniGene ID
Mm.208751
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004151.2 NP_001004151.1 1 144
GenBank mRNA AK047543.1 - 1 145
AY562225.1 - 1 98
BC138910.1 - 1 130 8861932
BC138912.1 - 1 130 8861934

View Full Product Details