Gene Transcripts

Gene Symbol
Ccdc37
Entrez Gene ID
243538
Gene Name
coiled-coil domain containing 37
Gene Aliases
AW048498, C030041G11Rik, C230069K22Rik
Chromosome Location
Chr. 6: 90403731 - 90430246 on Build GRCm38
UniGene ID
Mm.386876
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173775.3 NP_776136.2 14 1516
GenBank mRNA AK048790.1 - 6 2111
BC047211.1 - 12 1198 6392183
BC079632.1 - 2 244 5693354

View Full Product Details