Gene Transcripts

Gene Symbol
Usp35
Entrez Gene ID
244144
Gene Name
ubiquitin specific peptidase 35
Gene Aliases
Gm1088, Gm493
Chromosome Location
Chr. 7: 97309380 - 97332255 on Build GRCm38
UniGene ID
Mm.297541
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177412.1 NP_001170883.1 9 2507
GenBank mRNA BC059867.1 - 7 1210

View Full Product Details