Gene Transcripts
- Gene Symbol
- Avpr1b
- Entrez Gene ID
- 26361
- Gene Name
- arginine vasopressin receptor 1B
- Gene Aliases
- AVPR3, V3/V1b, VIBR, VPR3
- Chromosome Location
- Chr. 1: 131599114 - 131612000 on Build GRCm38
- UniGene ID
- Mm.89986
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011924.2 |
NP_036054.1
|
1 |
780 |
|
View Full Product Details