Gene Transcripts

Gene Symbol
Ccdc32
Entrez Gene ID
269336
Gene Name
coiled-coil domain containing 32
Gene Aliases
Gm631
Chromosome Location
Chr. 2: 119017779 - 119029406 on Build GRCm38
UniGene ID
Mm.292465
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199310.2 NP_955014.2 2 300
GenBank mRNA AK040670.1 - 2 270
AK132604.1 - 2 292
AK133813.1 - 2 274
AK147152.1 - 2 276
BC023926.1 - 2 294 5324154
BC052687.1 - 2 266
BY086759.1 - 2 301
BY262121.1 - 2 306

View Full Product Details