Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001037713.4 | NP_001032802.2 | 5 | 774 | |
NM_001291153.1 | NP_001278082.1 | 3 | 489 | ||
GenBank mRNA | BY726623.1 | - | 3 | 489 | |
CK130151.1 | - | 4 | 709 | ||
EL957551.1 | - | 3 | 479 |