Gene Transcripts

Gene Symbol
Serf2
Entrez Gene ID
378702
Gene Name
small EDRK-rich factor 2
Gene Aliases
C80083, Msmac1l, m4F5rel
Chromosome Location
Chr. 2: 121449198 - 121453420 on Build GRCm38
UniGene ID
Mm.262252
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011354.3 NP_035484.1 2, 3 353
GenBank mRNA AF073297.1 - 2, 3 134
AK002351.1 - 2, 3 129
AK008802.1 - 2, 3 142
AK010749.1 - 2, 3 185
AK012395.1 - 2, 3 146
AK012603.1 - 2, 3 132
AK018746.1 - 2, 3 129
AK151362.1 - 2, 3 146
AK168858.1 - 2, 3 133
AK169106.1 - 2, 3 135
BC081455.1 - 2, 3 137
BC092136.1 - 2, 3 166 6814342
CJ073462.1 - 2, 3 359

Gene Symbol
Fam221b
Entrez Gene ID
242408
Gene Name
family with sequence similarity 221, member B
Gene Aliases
4930412F15Rik
Chromosome Location
Chr. 4: 43659622 - 43668859 on Build GRCm38
UniGene ID
Mm.440423
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK038644.1 - 3 2931

View Full Product Details