Gene Transcripts

Gene Symbol
Ovol3
Entrez Gene ID
381867
Gene Name
OVO homolog-like 3 (Drosophila)
Gene Aliases
Gm1903, movo3
Chromosome Location
Chr. 7: 30233297 - 30237575 on Build GRCm38
UniGene ID
Mm.483986
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289817.1 NP_001276746.1 3 1636

View Full Product Details