Gene Transcripts

Gene Symbol
Scgb2b2
Entrez Gene ID
381970
Gene Name
secretoglobin, family 2B, member 2
Gene Aliases
Abpbg2, Abpe, lacrimal
Chromosome Location
Chr. 7: 31302769 - 31304977 on Build GRCm38
UniGene ID
Mm.333771
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207262.2 NP_997145.2 3 343
GenBank mRNA CK616217.1 - 3 343
CK616805.1 - 181

View Full Product Details