Gene Transcripts

Gene Symbol
Acpt
Entrez Gene ID
100503991
Gene Name
acid phosphatase, testicular
Gene Aliases
EG546967, Gm1432
Chromosome Location
Chr. 7: 44251761 - 44261396 on Build GRCm38
UniGene ID
Mm.367353
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195034.1 NP_001181963.1 3 235

View Full Product Details