Gene Transcripts

Gene Symbol
Arid3c
Entrez Gene ID
550619
Gene Name
AT rich interactive domain 3C (BRIGHT-like)
Gene Aliases
OTTMUSG00000006683
Chromosome Location
Chr. 4: 41723831 - 41731867 on Build GRCm38
UniGene ID
Mm.425179
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017362.2 NP_001017362.1 6 1022
NM_001252622.1 NP_001239551.1 6 1022

View Full Product Details