Gene Transcripts
- Gene Symbol
- Trim30e-ps1
- Entrez Gene ID
- 625321
- Gene Name
- tripartite motif-containing 30E, pseudogene 1
- Gene Aliases
- EG625321, Gm6577, Trim30-4
- Chromosome Location
- Chr. 7: 104532959 - 104535361 on Build GRCm38
- UniGene ID
- Mm.367236
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK140655.1 |
- |
3 |
867 |
|
BC147288.1 |
- |
3 |
328 |
9056358 |
View Full Product Details