Gene Transcripts

Gene Symbol
Apol11a
Entrez Gene ID
626615
Gene Name
apolipoprotein L 11a
Gene Aliases
EG626615
Chromosome Location
Chr. 15: 77508129 - 77517874 on Build GRCm38
UniGene ID
Mm.390820
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177533.1 NP_001171004.1 4 787

View Full Product Details