Gene Transcripts

Gene Symbol
Tas2r122
Entrez Gene ID
630845
Gene Name
taste receptor, type 2, member 122
Gene Aliases
T2R22, Tas2r22, mGR22
Chromosome Location
Chr. 6: 132710999 - 132711928 on Build GRCm38
UniGene ID
Mm.439643
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039128.1 NP_001034217.1 1 551

View Full Product Details