Gene Transcripts
- Gene Symbol
- Smpx
- Entrez Gene ID
- 66106
- Gene Name
- small muscle protein, X-linked
- Gene Aliases
- 1010001C09Rik, Csl
- Chromosome Location
- Chr. X: 157698973 - 157752591 on Build GRCm38
- UniGene ID
- Mm.140340
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252591.2 |
NP_001239520.1
|
2, 3 |
402 |
|
GenBank mRNA |
AA434782.1 |
- |
2, 3 |
257 |
|
AF364070.1 |
- |
2, 3 |
241 |
|
AI035961.1 |
- |
2, 3 |
216 |
|
AJ245772.1 |
- |
2, 3 |
257 |
|
AK003105.1 |
- |
2, 3 |
249 |
|
AK009857.1 |
- |
2, 3 |
249 |
|
AK010172.1 |
- |
2, 3 |
249 |
|
AY026524.1 |
- |
2, 3 |
248 |
|
BC051197.1 |
- |
2, 3 |
201 |
1397202 |
BC061051.1 |
- |
2, 3 |
163 |
30300606 |
BC132358.1 |
- |
2, 3 |
77 |
40130635 |
BC138801.1 |
- |
2, 3 |
77 |
8861823 |
View Full Product Details