Gene Transcripts

Gene Symbol
Uggt2
Entrez Gene ID
66435
Gene Name
UDP-glucose glycoprotein glucosyltransferase 2
Gene Aliases
1810064L21Rik, 3110001A05Rik, 3110027P15Rik, A230065J02Rik, AW047562, Ugcgl2
Chromosome Location
Chr. 14: 118981765 - 119099437 on Build GRCm38
UniGene ID
Mm.213406
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081252.2 NP_001074721.2 9 1046
GenBank mRNA AK038817.1 - 9 1042
AK048685.1 - 9 1051
AK052077.1 - 9 1046

View Full Product Details