Gene Transcripts

Gene Symbol
Pomp
Entrez Gene ID
66537
Gene Name
proteasome maturation protein
Gene Aliases
2510048O06Rik, Ump1
Chromosome Location
Chr. 5: 147860628 - 147875778 on Build GRCm38
UniGene ID
Mm.332855
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025624.2 NP_079900.1 4 298
GenBank mRNA AK003439.1 - 4 291
AK011115.1 - 4 299
AK012455.1 - 4 260
AK013934.1 - 4 292
AK014296.1 - 4 262
AK044283.1 - 4 274
AK132041.1 - 275
AK167958.1 - 4 299
BC027016.1 - 4 289 5375452

View Full Product Details