Gene Transcripts

Gene Symbol
Pex10
Entrez Gene ID
668173
Gene Name
peroxisomal biogenesis factor 10
Gene Aliases
AV128229, Gm142
Chromosome Location
Chr. 4: 155067030 - 155072406 on Build GRCm38
UniGene ID
Mm.133114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042407.1 NP_001035866.1 3 565
GenBank mRNA AK043682.1 - 3 539
BC147304.1 - 3 550 9056372
BC147305.1 - 3 550 9056374
CB203398.1 - 2 139
CJ064027.1 - 3 539

View Full Product Details