Gene Transcripts

Gene Symbol
Ttc9c
Entrez Gene ID
70387
Gene Name
tetratricopeptide repeat domain 9C
Gene Aliases
2210019E14Rik, 6330408J23Rik
Chromosome Location
Chr. 19: 8809066 - 8819780 on Build GRCm38
UniGene ID
Mm.261082
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027412.3 NP_081688.2 3, 4 703
GenBank mRNA AK008753.1 - 2, 3 1034
AK018145.1 - 3, 4 743
AK169288.1 - 3, 4 704
AK169296.1 - 3, 4 704
BC035304.1 - 2, 3 614
BC043061.1 - 2, 3 1015 5704924
BC058967.1 - 2, 3 1015 5704924

View Full Product Details