Gene Transcripts

Gene Symbol
Atp13a2
Entrez Gene ID
74772
Gene Name
ATPase type 13A2
Gene Aliases
1110012E06Rik, AA589443
Chromosome Location
Chr. 4: 140986865 - 141007701 on Build GRCm38
UniGene ID
Mm.205625
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK156104.1 - 15 1592
AK194090.1 - 1 84
AK197762.1 - 1 84
BC023746.1 - 15 1588 5353766
BC042661.1 - 15 1558 4192094

View Full Product Details