Gene Transcripts

Gene Symbol
Avl9
Entrez Gene ID
78937
Gene Name
AVL9 homolog (S. cerevisiase)
Gene Aliases
5830411G16Rik, D730049P16Rik, mKIAA0241
Chromosome Location
Chr. 6: 56714905 - 56761912 on Build GRCm38
UniGene ID
Mm.105343
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030235.1 NP_084511.1 13 1880
GenBank mRNA AK020003.1 - 13 1857
AK083605.1 - 13 1837
AK122228.1 - 13 1858
AK147365.1 - 13 1882
BC117878.1 - 13 1842 40095961
BC117879.1 - 13 1842

View Full Product Details