Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_139229.4 | NP_631975.3 | 3 | 1238 | |
GenBank mRNA | AB041610.2 | - | 3 | 1161 | |
AK049957.1 | - | 3 | 1186 | ||
AK083734.1 | - | 3 | 1180 | ||
AK145515.1 | - | 3 | 1191 | ||
AK145727.1 | - | 3 | 1182 | ||
AK153617.1 | - | 3 | 1182 | ||
AK156186.1 | - | 3 | 1181 | ||
AK159569.1 | - | 3 | 1179 | ||
BC094654.1 | - | 1174 | 6853777 |