Gene Transcripts

Gene Symbol
Acsm2a
Entrez Gene ID
246263
Gene Name
acyl-CoA synthetase medium-chain family member 2A
Gene Aliases
Acsm2, KS
Chromosome Location
Chr. 1: 189289934 - 189329011 on Build Rnor_6.0
UniGene ID
Rn.14875
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144748.1 NP_653349.1 11 1493
XM_008759677.1 XP_008757899.1 11 1478
XM_008759678.1 XP_008757900.1 10 1434
XM_008759679.1 XP_008757901.1 8 1231
GenBank mRNA AF062389.1 - 11 1493
BC078721.1 - 11 1470 7110246

View Full Product Details