Gene Transcripts

Gene Symbol
B3gnt4
Entrez Gene ID
288752
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
Gene Aliases
Chromosome Location
Chr. 12: 38502987 - 38506793 on Build Rnor_6.0
UniGene ID
Rn.218458
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105938.1 NP_001099408.1 1 556
XM_006249335.2 XP_006249397.1 2 747
XM_006249336.2 XP_006249398.1 2 523

View Full Product Details