Gene Transcripts

Gene Symbol
Tmc3
Entrez Gene ID
293065
Gene Name
transmembrane channel-like 3
Gene Aliases
Stard5
Chromosome Location
Chr. 1: 145715829 - 145762512 on Build Rnor_6.0
UniGene ID
Rn.138440
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170432.1 NP_001163903.1 22 3087
XM_008759610.1 XP_008757832.1 23 3314

View Full Product Details