Gene Transcripts

Gene Symbol
Secisbp2l
Entrez Gene ID
296115
Gene Name
SECIS binding protein 2-like
Gene Aliases
RGD1559930
Chromosome Location
Chr. 3: 118005656 - 118051403 on Build Rnor_6.0
UniGene ID
Rn.48946
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168527.1 NP_001161999.1 5, 6 1132
XM_008762152.1 XP_008760374.1 5, 6 1089

View Full Product Details