Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001011974.1 | NP_001011974.1 | 2 | 1541 | |
XM_006238184.2 | XP_006238246.1 | 2 | 2106 | ||
XM_006238185.2 | XP_006238247.1 | 2 | 1523 | ||
XM_006238186.2 | XP_006238248.1 | 2 | 1622 | ||
XM_008763733.1 | XP_008761955.1 | 3 | 1588 | ||
GenBank mRNA | BC085790.1 | - | 2 | 1541 |