Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007655.1 | NP_001007656.1 | 3, 4 | 609 | |
XM_006239554.2 | XP_006239616.1 | 3, 4 | 461 | ||
XM_006239555.2 | XP_006239617.1 | 4, 5 | 796 | ||
XM_006239557.2 | XP_006239619.1 | 2, 3 | 456 | ||
XM_006239558.2 | XP_006239620.1 | 2, 3 | 298 | ||
XM_008764352.1 | XP_008762574.1 | 3, 4 | 619 | ||
XM_008764353.1 | XP_008762575.1 | 3, 4 | 1689 | ||
XM_008764354.1 | XP_008762576.1 | 4, 5 | 800 | ||
XM_008764355.1 | XP_008762577.1 | 4, 5 | 1745 | ||
GenBank mRNA | BC079115.1 | - | 3, 4 | 609 |