Gene Transcripts

Gene Symbol
Trappc6b
Entrez Gene ID
299075
Gene Name
trafficking protein particle complex 6B
Gene Aliases
RGD1309325
Chromosome Location
Chr. 6: 80142126 - 80153216 on Build Rnor_6.0
UniGene ID
Rn.22788
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106733.1 NP_001100203.1 2, 3 317
GenBank mRNA FQ230024.1 - 2, 3 80

View Full Product Details