Gene Transcripts

Gene Symbol
Txndc11
Entrez Gene ID
302899
Gene Name
thioredoxin domain containing 11
Gene Aliases
RGD1307538
Chromosome Location
Chr. 10: 4578469 - 4635963 on Build Rnor_6.0
UniGene ID
Rn.54595
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127532.1 NP_001121004.1 4 751
XM_008767481.1 XP_008765703.1 3 957
GenBank mRNA BC166713.1 - 4 751

View Full Product Details