Gene Transcripts

Gene Symbol
Zscan10
Entrez Gene ID
302962
Gene Name
zinc finger and SCAN domain containing 10
Gene Aliases
Zfp206, Znf206
Chromosome Location
Chr. 10: 12929471 - 12939439 on Build Rnor_6.0
UniGene ID
Rn.106364
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106981.1 NP_001100451.1 3 745
XM_006245870.2 XP_006245932.1 3 745

View Full Product Details