Gene Transcripts

Gene Symbol
Atg16l2
Entrez Gene ID
308865
Gene Name
autophagy related 16-like 2 (S. cerevisiae)
Gene Aliases
RGD1311400
Chromosome Location
Chr. 1: 166401271 - 166415076 on Build Rnor_6.0
UniGene ID
Rn.104199
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191560.1 NP_001178489.1 14 1392
XM_006229876.1 XP_006229938.1 13 2841
XM_006229877.2 XP_006229939.1 12 1422

View Full Product Details