Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001107896.1 | NP_001101366.1 | 6, 7 | 770 | |
XM_008763382.1 | XP_008761604.1 | 8, 9 | 1695 | ||
XM_008763383.1 | XP_008761605.1 | 7, 8 | 1316 | ||
XM_008763384.1 | XP_008761606.1 | 7, 8 | 1576 | ||
XM_008763385.1 | XP_008761607.1 | 7, 8 | 1576 | ||
XM_008763386.1 | XP_008761608.1 | 7, 8 | 1576 | ||
XM_008763387.1 | XP_008761609.1 | 7, 8 | 1576 | ||
XM_008763388.1 | XP_008761610.1 | 7, 8 | 1576 | ||
XM_008763389.1 | XP_008761611.1 | 7, 8 | 1576 | ||
XM_008763390.1 | XP_008761612.1 | 7, 8 | 1576 | ||
XM_008763391.1 | XP_008761613.1 | 7, 8 | 1576 | ||
XM_008763392.1 | XP_008761614.1 | 7, 8 | 1576 | ||
XM_008763393.1 | XP_008761615.1 | 7, 8 | 1576 |