Gene Transcripts

Gene Symbol
Chrdl1
Entrez Gene ID
363455
Gene Name
chordin-like 1
Gene Aliases
Kjr, Neurogenesin-1, Ng1
Chromosome Location
Chr. X: 114554359 - 114658960 on Build Rnor_6.0
UniGene ID
Rn.211154
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199502.1 NP_955796.1 4 692
XM_006257373.2 XP_006257435.1 4 716
GenBank mRNA AB080636.1 - 4 692

View Full Product Details