Gene Transcripts

Gene Symbol
Adam1a
Entrez Gene ID
56777
Gene Name
a disintegrin and metallopeptidase domain 1a
Gene Aliases
Adam1, PH-30a
Chromosome Location
Chr. 12: 40533731 - 40536632 on Build Rnor_6.0
UniGene ID
Rn.42919
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020078.1 NP_064463.1 1 718
GenBank mRNA BC081807.1 - 1 817
Y08616.1 - 1 718

Gene Symbol
LOC100360238
Entrez Gene ID
100360238
Gene Name
rCG21419-like
Gene Aliases
Chromosome Location
Chr. 12: 40551792 - 40556984 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751190.3 XP_003751238.1 1 2323

View Full Product Details