Gene Transcripts

Gene Symbol
C1qtnf5
Entrez Gene ID
315598
Gene Name
C1q and tumor necrosis factor related protein 5
Gene Aliases
Chromosome Location
Chr. 8: 48443767 - 48445637 on Build Rnor_6.0
UniGene ID
Rn.99884
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012123.1 NP_001012123.1 3 635
GenBank mRNA BC089992.1 - 3 635

View Full Product Details