Gene Transcripts

Gene Symbol
Aqp12a
Entrez Gene ID
367316
Gene Name
aquaporin 12A
Gene Aliases
Aqp12, Aqp12b, RGD1562727
Chromosome Location
Chr. 9: 100156154 - 100166218 on Build Rnor_6.0
UniGene ID
Rn.20532
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109009.1 NP_001102479.1 3 852

View Full Product Details