Gene Transcripts

Gene Symbol
Slc4a3
Entrez Gene ID
24781
Gene Name
solute carrier family 4 (anion exchanger), member 3
Gene Aliases
Ae3, Aep3
Chromosome Location
Chr. 9: 82743028 - 82758213 on Build Rnor_6.0
UniGene ID
Rn.87739
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017049.1 NP_058745.1 21 3528
XM_006245149.2 XP_006245211.1 17 3587
GenBank mRNA J05167.1 - 21 3528

View Full Product Details