Gene Transcripts

Gene Symbol
Rab12
Entrez Gene ID
25530
Gene Name
RAB12, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 9: 114683985 - 114709613 on Build Rnor_6.0
UniGene ID
Rn.11021
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013017.1 NP_037149.1 252
GenBank mRNA CB692926.1 - 1 252
M83676.1 - 1 47

View Full Product Details