Gene Transcripts

Gene Symbol
Ranbp3l
Entrez Gene ID
294789
Gene Name
RAN binding protein 3-like
Gene Aliases
RGD1307559
Chromosome Location
Chr. 2: 58221457 - 58235333 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006232030.2 XP_006232092.1 593
XM_006232031.2 XP_006232093.1 592

View Full Product Details