Gene Transcripts

Gene Symbol
Apol11a
Entrez Gene ID
300060
Gene Name
apolipoprotein L 11a
Gene Aliases
Apol11a-ps1, RGD1309798
Chromosome Location
Chr. 7: 118835316 - 118858333 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006241949.2 XP_006242011.1 4 482
XM_008765647.1 XP_008763869.1 3 410

View Full Product Details