Gene Transcripts

Gene Symbol
Ubxn7
Entrez Gene ID
303878
Gene Name
UBX domain protein 7
Gene Aliases
RGD1306346, Ubxd7
Chromosome Location
Chr. 11: 71662786 - 71695031 on Build Rnor_6.0
UniGene ID
Rn.230486
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107086.1 NP_001100556.1 3 339

View Full Product Details